Specifications
- Description:Quick Connect™ jacket adaptor with injection cap, 23 G pin
- Cat. no.:89134-212
- Supplier No.:QCJA-23
Specifications
About this item
The Quick Connect™ Jacket Adaptor is designed for researchers who prefer a woven rodent jacket over an elastomeric harness for continuous or intermittent infusion. The Jacket Adaptor utilizes the same luer design as the Quick Connect™ Harness, but connects securely to a cloth jacket with Velcro™, and can be adjusted as the animal grows.
- Attached injection cap can be removed, or pierced with a Huber needle
- Choose from two catheter connector pin sizes to fit 21G and 23G catheters
- To be used with an SAI/Lomir Jacket
- Fit can be adjusted as the animal grows
- Individually packaged
- EtO exposed
These jackets are part of SAI's Quick Connect™ System, which includes the Quick Connect™ Tether, Harness, and Swivel for tethered small animal infusion studies.