Distilling Heads, Chemglass
Supplier: Chemglass
Certificates
About this item
Simple distillation head with a condenser sealed to the adapter at a 75° angle.
- The sidearm is sealed at a height for use with a 1" immersion thermometer
- The 24/40 and 29/42 sizes are made to accept a 10/30, 3" immersion thermometer
The inner drip tip joint on the condenser and inner joint on the uptake tube are the same size. The top outer joint on the 14/20 size can be converted to a 10/18 thermometer joint by using a bushing adapter.
Customers Who Bought This Also Bought