Order Entry
ContactUsLinkComponent
Drying Apparatus, Abderhalden, Chemglass
Drying Apparatus, Abderhalden, Chemglass
Catalog #: 80067-960
Supplier:  Chemglass
Small Business Enterprise
DRYING APPARATUS, ABDERHALDEN
Drying Apparatus, Abderhalden, Chemglass
Catalog #: 80067-960
Supplier:  Chemglass
Small Business Enterprise

Specifications

  • Width:
    350 mm
  • Description:
    Drying Apparatus, Abderhalden, Complete
  • Height:
    740 mm
  • Cat. no.:
    80067-960
  • Supplier No.:
    CG-1289-01

Specifications

About this item

Desiccator glasses feature round bottoms.

  • Abderhalden style
  • For drying samples at a temperature
  • As a function of the solvent boiling point

The desiccant flask has a 34/45 outer joint for connection to the drying chamber. All other joints are 24/40.

Delivery: Complete apparatus consists of a desiccant flask, drying chamber, round bottom flask and Allihn condenser.
Customers Who Bought This Also Bought