Jacketed Filter Reactor Systems, 10 L, Electric Motor, Chemglass
Catalog #: 89209-352
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:Jacketed Cylindrical Process Filter Reactor System Complete
- Motor Type:Electric motor
- Cat. no.:89209-352
- Supplier No.:CG-1959-F610
Specifications
About this item
Jacketed filter glass reactors feature single or multi-step.
- Complete reactor system on open air, auto center stands
- Maximum temperature of 150 °C limit for filter base
- Filter base eliminates material transfers and significantly reduces losses
Can be used for single or multi-step reactions and filtrations in the same vessel, production and process scale peptide synthesis.
Ordering information: Filter reactors are for vacuum/gravity filtration use only.
Customers Who Bought This Also Bought