Specifications
- Length:12"
- Description:QuickConnect™ Dual Luer Spring Tether
- Cat. no.:89134-250
- Supplier No.:QCT-12
Specifications
About this item
The Dual Luer Spring Tether is a standard, stainless steel tether with a male luered polyurethane extension line built inside. The male luered fittings connect to the QuickConnect™ Harness and QuickConnect™ Swivel to complete the system.
- Connections at the catheter and swivel ends are fast and easy
- Compatible with any female Luer
- Lined with polyurethane tubing
- Length is customizable
This flexible stainless steel spring tether will protect the infusion line from damage due to biting during both short-term and long-term infusion studies. Easily connect to a QuickConnect™ swivel for freedom of movement without any tether twisting.