ContactUsLinkComponent
Multi-Directional Swivel Mount Assembly, SAI Infusion Technologies
Multi-Directional Swivel Mount Assembly, SAI Infusion Technologies
Catalog #: 89134-254
QUICK CONNECT SWIVEL MOUNT
Multi-Directional Swivel Mount Assembly, SAI Infusion Technologies
Catalog #: 89134-254
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Multi-Directional Swivel Mount Assembly, with Gimbal Mount and Adjustable 8" Post
  • Cat. no.:
    89134-254
  • Supplier No.:
    GMSHK
Specifications
About this item
Completely adjustable, multi-directional swivel mounting system made of anodized aluminum.
  • Reduce animal stress and tether tangles with the gimbal mount's unparalleled 4-axes of movement
  • Stainless steel base quickly attaches to any style caging and is cage washer-resistant
  • Quick-release swivel holder allows for continuous infusion during body weights and cage changes
  • Durable anodized aluminum components
The swivel is easily raised and lowered with a thumb screw, and the 8" post can be trimmed to any height. Interchangeable fittings allow use of the multi-directional swivel holder with or without the counter balance arm.
Caution: This product is for laboratory animal use only.
Customers Who Bought This Also Bought