About this item
Tapered polyurethane rat arterial catheters are designed to enter the smaller, more muscular vessels while maintaining strength and durability at the externalization point.
- Tapered design improves fluid dynamics
- Suture beads secure the catheter for improved patency
- Small tip size and smooth surface improve the catheter implantation process
- Radio opaque for catheter placement verification
- Accessible with a 22G blunt needle
Both a tapered and a step-up option of this device are available.
For animal 150 g plus.
Tip O.D 2.3F, tip length 4 cm