Flasks, Heavy Wall, Round Bottom, Single Neck With Hooks, Chemglass
Supplier: Avantor
Certificates
About this item
Round bottom, heavy wall single short neck flask with a standard taper outer joint.
- High-quality heavy wall flasks are short neck
- Flasks feature a groove to aid in clamping of the flask
Flask has two hooks 180° apart for use with CG-110 stainless steel springs.
Customers Who Bought This Also Bought