Order Entry
ContactUsLinkComponent
Mouse Recombinant R-Spondin-1 (from CHO cells)
Mouse Recombinant R-Spondin-1 (from CHO cells)
Catalog #: 76303-960
Supplier:  Peprotech
CAS Number:  
Mouse Recombinant R-Spondin-1 (from CHO cells)
Catalog #: 76303-960
Supplier:  Peprotech
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Gene ID:
    192199
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Conjugation:
    Unconjugated
  • Size:
    50 µg
  • Biological activity:
    R-Spondin-1 enhances BMP-2-mediated differentiation of MC3T3-E1 cells.
  • Protein synonyms:
    Roof plate-specific Spondin-1|RSPO1
  • Protein/peptide name:
    R-Spondin-1
  • Purity:
    95%
  • Molecular weight:
    27 kDa
  • Sequence:
    SRGIKGKRQR RISAEGSQAC AKGCELCSEV NGCLKCSPKL FILLERNDIR QVGVCLPSCP PGYFDARNPD MNKCIKCKIE HCEACFSHNF CTKCQEGLYL HKGRCYPACP EGSTAANSTM ECGSPAQCEM SEWSPWGPCS KKRKLCGFRK GSEERTRRVL HAPGGDHTTC SDTKETRKCT VRRTPCPEGQ KRRKGGQGRR ENANRHPARK NSKEPGSNSR RHKGQQQPQP GTTGPLTSVG PTWAQ
  • Endotoxin level:
    low
  • Formulation:
    Lyophilized
  • Shipping temperature:
    Ambient
  • Cat. no.:
    76303-960

Specifications

About this item

R-Spondin-1 (Rspo-1) belongs to the (Rspo) family of Wnt modulators. Currently, the family consists of four structurally related secreted ligands (Rspo 1-4), all containing the furin-like and thrombospondin structural domains. Rspo-1 is expressed in certain areas of the developing central nervous system, as well as in the adrenal glands, ovary, testis, thyroid, and trachea. Rspo can interact with the Frizzled/LRP6 receptor complex in a manner that stimulates the Wnt/beta-catenin signaling pathway. Recombinant Murine R-Spondin-1 is a 27.1 kDa protein consisting of 245 amino acid residues. Due to glycosylation, R-Spondin-1 migrates at an apparent molecular weight of approximately 40.0 kDa by SDS-PAGE analysis under reducing conditions.

Customers Who Bought This Also Bought