Order Entry
ContactUsLinkComponent
Human Recombinant IL-22 (from E. coli)
Human Recombinant IL-22 (from E. coli)
Catalog #: 76303-794
Supplier:  Peprotech
CAS Number:  
Human Recombinant IL-22 (from E. coli)
Catalog #: 76303-794
Supplier:  Peprotech
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Animal-free:
    Y
  • Gene ID:
    50616
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Conjugation:
    Unconjugated
  • Size:
    50 µg
  • Biological activity:
    Assay #1: Determined by its ability to activate STAT following receptor ligand interaction. Assay #2: Measured by its ability to induce IL-10 secretion in COLO 205 (human colon carcinoma cells). The expected ED50 for this effect is 0.7- 1.0 ng/ml.
  • Protein synonyms:
    IL-TIF|Cytokine ZCYTO18|Interleukin-22
  • Protein/peptide name:
    IL-22
  • Purity:
    98%
  • Molecular weight:
    33.6 kDa
  • Sequence:
    MAPISSHCRL DKSNFQQPYI TNRTFMLAKE ASLADNNTDV RLIGEKLFHG VSMSERCYLM KQVLNFTLEE VLFPQSDRFQ PYMQEVVPFL ARLSNRLSTC HIEGDDLHIQ RNVQKLKDTV KKLGESGEIK AIGELDLLFM SLRNACI
  • Endotoxin level:
    low
  • Formulation:
    Lyophilized
  • Shipping temperature:
    Ambient
  • Cat. no.:
    76303-794

Specifications

About this item

IL-22 is a member of the IL-10 family of regulatory cytokines, which includes IL-10, IL-19, IL-20, IL-22, IL-24 and IL-26. Members of this family share partial homology in their amino acid sequences, but they are dissimilar in their biological functions. Produced by T lymphocytes, IL-22 inhibits IL-4 production by Th2 cells, and induces acute phase reactants in the liver and pancreas. IL-22 signals through a receptor system consisting of IL-10Rbeta/CRF2-4 and IL-22R, both of which are members of the class II cytokine-receptor family. Recombinant Human IL-22 is a 33.6 kDa, non-disulfide-linked, homodimeric protein consisting of two 147 amino acid polypeptide chains.

Customers Who Bought This Also Bought