Order Entry
ContactUsLinkComponent
Human Recombinant BAFF (from E. coli)
Human Recombinant BAFF (from E. coli)
Catalog #: 76303-908
Supplier:  Peprotech
CAS Number:  
Human Recombinant BAFF (from E. coli)
Catalog #: 76303-908
Supplier:  Peprotech
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Animal-free:
    Y
  • Gene ID:
    10673
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Conjugation:
    Unconjugated
  • Size:
    50 µg
  • Biological activity:
    Determined by a mouse splenocyte survival assay. The expected ED50 for this effect is 0.5-2.0 ug/ml.
  • Protein synonyms:
    THANK|ZNTF4|TALL-1|TNFSF20|BLyS|TNFSF13B|B cell Activating Factor belonging to the TNF family
  • Protein/peptide name:
    BAFF
  • Purity:
    95%
  • Molecular weight:
    17 kDa
  • Sequence:
    AVQGPEETVT QDCLQLIADS ETPTIQKGSY TFVPWLLSFK RGSALEEKEN KILVKETGYF FIYGQVLYTD KTYAMGHLIQ RKKVHVFGDE LSLVTLFRCI QNMPETLPNN SCYSAGIAKL EEGDELQLAI PRENAQISLD GDVTFFGALK LL
  • Endotoxin level:
    low
  • Formulation:
    Lyophilized
  • Shipping temperature:
    Ambient
  • Cat. no.:
    76303-908

Specifications

About this item

BAFF, a member of the TNF family of ligands, is expressed in T cells, macrophages, monocytes and dendritic cells. BAFF is involved in stimulation of B and T cell function, and is an important survival and maturation factor for peripheral B cells. BAFF signals through three different TNF receptors, TACI, BCMA and BAFFR. The human BAFF gene codes for a 285 amino acid type II transmembrane protein containing a 46 amino acid cytoplasmic domain, a 21 amino acid transmembrane domain, and a 218 amino acid extracellular domain. Recombinant Human soluble BAFF is a 152 amino acid polypeptide (17.0 kDa), which contains the TNF-like portion of the extracellular domain of BAFF.

Customers Who Bought This Also Bought