Order Entry
ContactUsLinkComponent
Human Recombinant PDGF-BB (from E. coli)
Human Recombinant PDGF-BB (from E. coli)
Catalog #: 76303-584
Supplier:  Peprotech
CAS Number:  
Human Recombinant PDGF-BB (from E. coli)
Catalog #: 76303-584
Supplier:  Peprotech
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Animal-free:
    Y
  • Gene ID:
    5155
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Conjugation:
    Unconjugated
  • Size:
    50 µg
  • Biological activity:
    Determined by the dose-dependent stimulation of the proliferation of Balb/c 3T3 cells. The expected ED50 for this effect is 1.0-3.0 ng/ml.
  • Protein synonyms:
    Osteosarcoma-derived Growth Factor (ODGF)|Platelet-Derived Growth Factor-BB|Glioma-derived growth factor (GDGF)
  • Protein/peptide name:
    PDGF-BB
  • Purity:
    98%
  • Molecular weight:
    24.3 kDa
  • Sequence:
    SLGSLTIAEP AMIAECKTRT EVFEISRRLI DRTNANFLVW PPCVEVQRCS GCCNNRNVQC RPTQVQLRPV QVRKIEIVRK KPIFKKATVT LEDHLACKCE TVAAARPVT
  • Endotoxin level:
    low
  • Formulation:
    Lyophilized
  • Shipping temperature:
    Ambient
  • Cat. no.:
    76303-584

Specifications

About this item

  • Animal-free

PDGFs are disulfide-linked dimers consisting of two 12.0-13.5 kDa polypeptide chains, designated PDGF-A and PDGF-B chains. The three naturally occurring PDGFs, PDGF-AA, PDGF-BB and PDGF-AB, are potent mitogens for a variety of cell types, including smooth muscle cells, connective tissue cells, bone and cartilage cells, and some blood cells. The PDGFs are stored in platelet alpha-granules, and are released upon platelet activation. The PDGFs are involved in a number of biological processes, including hyperplasia, chemotaxis, embryonic neuron development, and respiratory tubule epithelial cell development. Two distinct signaling receptors used by PDGFs have been identified and named PDGFR-alpha and PDGFR-beta. PDGFR-alpha is a high-affinity receptor for each of the three PDGF forms. On the other hand, PDGFR-beta interacts with only PDGF-BB and PDGF-AB. Recombinant Human PDGF-BB is a 24.3 kDa disulfide-linked homodimer of two beta chains (218 total amino acids).

Customers Who Bought This Also Bought