About this item
High performance fume hoods with integrated sash sensors for automatic multi-speed blower control.
- High performance chemical fume hoods
- Integrated sash sensors for blower speed control
- Factory-installed Guardian™ digital airflow monitor
- One-piece molded fiberglass liner
Protector® Premier® I-S™ laboratory hoods incorporate a sleek interior with a molded one-piece fiberglass liner, the signature feature of leading line of fume hoods since 1961. The one-piece liner of specially-formulated, fiberglass-reinforced polyester offers corrosion and fire resistance and is easy to cleanup. Without seams, the interior has fewer points of deterioration for longer life.
Designed for use with Intelli-Sense multi-speed blowers, Protector® Premier® I-S™ hoods feature integrated sash position blower controls. As the sash is raised or lowered, sensors alert the blower to change speeds to maintain a safe face velocity.
Protector® Premier® hoods incorporate many containment-enhancing features including Clean-Sweep™ technology and the Eco-Foil™ air foil. Testing confirms the Protector® Premier® I-S™ hoods meets the SEFA-1 standard of a low velocity, high performance hood and may be operated as low as 60 fpm.
Specifications
- Catalog No:
- 76330-538
- 76330-098
- 76330-536
- 76330-096
- 76330-102
- 76330-100
- 76330-540
- 76330-380
- Environmentally Preferable:
- Sustainable Packaging, Energy Efficient
- Sustainable Packaging, Energy Efficient
- Sustainable Packaging, Energy Efficient
- Sustainable Packaging, Energy Efficient
- Sustainable Packaging, Energy Efficient
- Sustainable Packaging, Energy Efficient
- Sustainable Packaging, Energy Efficient
- Sustainable Packaging, Energy Efficient