Metamorphic Rock Collection
Catalog #: 470315-514
Supplier: Avantor
Specifications
- Description:Metamorphic Rock Collection
- No. of Samples:12
- Cat. no.:470315-514
Specifications
About this item
Perfect for introductory lessons on metamorphic rocks.
- Hand cut by Ward's geologists
- Specimen list with localities included
Study textures, grain sizes, and other features altered by pressure, temperature, and other natural forces, characteristic of metamorphic rocks. The set is housed in a compartmented collection box and comes with an identification list.
Contents: 12 numbered samples, 1×2"
Rocks: gneiss (2), marble (2), quartzite, schist (3), phyllite, slate (2), amphibolite
Customers Who Bought This Also Bought