Fracture Collection
Catalog #: 470315-516
Supplier: Avantor
Specifications
- Description:Fracture collection
- No. of Samples:6
- Cat. no.:470315-516
Specifications
About this item
Demonstrates irregular breakage of minerals.
Each specimen illustrates a type of mineral breakage that does not leave a smooth cleavage surface. The set is housed in a compartmented collection box, and an identification list is included.
Contents: 6 numbered samples, 1×1"
Even Fracture - Halite
Uneven Fracture - Agate
Conchoidal Fracture - Obsidian
Subconchoidal Fracture - Rose Quartz
Splintery Fracture - Satin Spar Gypsum
Hackly Fracture - Copper
Customers Who Bought This Also Bought