Classroom Mineral Collection
Catalog #: 470315-612
Supplier: Avantor
Specifications
- Description:Classroom Mineral Collection
- No. of Samples:36
- Cat. no.:470315-612
Specifications
About this item
A fascinating and colorful classroom study set.
The mineral collection containing mineral collected from all around the world is ideal for introducing students to Mohs' hardness testing, basic rock-forming minerals and minerals and ores used in products important to our economy. The set is housed in a compartment box and comes with an identification sheet with localities and guide book to rocks and minerals.
Customers Who Bought This Also Bought