Stomacher® Bag Clips, Seward
Catalog #: 97035-482
Supplier: Seward Laboratory System
Specifications
- Capacity:80 - 400 ml
- Description:Bag Clips
- Cat. no.:97035-482
- Supplier No.:BA6099
Specifications
About this item
Stomacher® Bag Clips allow fast and easy securing for all 80 to 400 ml sample bags.
- Ideal for short term storage of samples
- Eliminates any leakage during storage or incubation
- Provides a reusable alternative to closure bags
- Easily cleaned with most lab disinfectants (not autoclavable)
A full range of Stomacher® accessories to help provide a safe and efficient working environment in your laboratory.
Customers Who Bought This Also Bought