About this item
Flatpack Electroporation Chambers offer higher sample volume capacity than standard electroporation cuvettes of similar gap distances.
- Sterile
- Aluminium
- Convenient single use format
- Compatible with BTX safety stands
Flatpack (Model 486) Chambers feature unique flow-through design with a 0.56 mm gap and volume of 10 to 85 ul. They provide field strengths as high as 40 kV/cm, ideal for high efficiency transformation of bacteria.
Flatpack (Model 485 Chambers) feature a non-flow-through design, 1.83 mm gap and maximum fill volume of 1.5 ml. They are appropriate for either prokaryotic or eukaryotic electroporation applications - transfecting or transforming a variety of organisms in vitro such as bacteria, mammalian cells, yeast, plant protoplasts and more.
Generator compatibility: Gemini X2, ECM 630, ECM 830, Legacy ECM 2001, ECM 2001+.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 58017-927
- 58017-929
- Model:
- 486
- 485