TX-750 Swinging Bucket Rotor Packages, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Increase lab productivity with the Thermo Scientific™ TX-750 4×750ml Swinging Bucket Rotor packages, delivering maximum flexibility at 4700 rpm.
Auto-Lock™ III Rotor System allows easy push-button installation and exchange of rotors, as well as easy access to the rotor chamber for quick cleaning.
Delivery: All Packages Include: TX-750 rotor cross, TX-750 buckets (set of 4), ClickSeal biocontainment lids (set of 4)
Cell culture package includes 50 ml adapter (set of 4), 15 ml adapter (set of 4)
Blood tube package includes: 5/7 ml adapter (set of 4), 10 ml adapter (set of 4)
Cell culture package includes 50 ml adapter (set of 4), 15 ml adapter (set of 4)
Blood tube package includes: 5/7 ml adapter (set of 4), 10 ml adapter (set of 4)
Customers Who Bought This Also Bought