Ward's® Essentials Standard Compound Microscope
Catalog #: 470315-506
Supplier: VWR
Specifications
- Description:Compound Monocular Standard Microscope
- Cat. no.:470315-506
- Illumination:Rechargeable LED
Specifications
About this item
Ideal for biological use and educational applications.
- 40×, 100× and 400× magnification
- Rechargeable LED illumination
- Inclined rotatable head
- 1 year limited warranty
This microscope model includes the most commonly specified features for standard high school instruction. The glass-coated DIN optics provide clear, crisp images and the rechargeable LED illumination offers added versatility and portability.
Customers Who Bought This Also Bought