Celestron Telescope Eyepiece and Filter Kit, 1.25"
Catalog #: 470136-650
Supplier: Celestron International
Specifications
- Magnification:3, 4, 6, 6.5, 8, 8.5, 13, 16, 17 and 32 mm
- Description:Eyepiece and filter kit, 1.25"
- Cat. no.:470136-650
- Supplier No.:94303
Specifications
About this item
This 1.25" eyepiece and filter kit includes 14 accessories that will add new life to your telescope and your observing!
- The most popular planetary filters are included in this kit
Have fun seeing what a blue filter will reveal about jupiter or unveil the polar ice caps on mars with a red or orange filter. A moon filter will increase your enjoyment of lunar observations by dimming the brightness and increasing contrast.
Delivery: You’ll get five quality Plossl eyepieces, a 2X Barlow Lens, six colored filters to improve observations of the planets, a Moon filter, and a foam-fitted aluminum carry case with extra room to grow. Five eyepieces and a Barlow lens equals 10 different magnifications to choose from, including the equivalent of a 3, 4, 6, 6.5, 8, 8.5, 13, 16, 17 and 32 mm. And that doesn’t include eyepieces you may already have in your collection, which will now achieve double the magnification with the addition of the Barlow.
Customers Who Bought This Also Bought