TLC Plates, Special Layers, Glass Backing, Chiral Plate, Macherey-Nagel
Catalog #: 29444-524
Supplier: Macherey-Nagel
Small Business Enterprise
Specifications
- Coating material:Chiral impregnated silica gel
- Layer thickness:250 µm
- Plate Size:100×100 mm
- Description:Chiral Plate
- Cat. no.:29444-524
- Fluorescent Indicator:UV₂₅₄
- Supplier No.:811059
Specifications
About this item
These glass plates with a special layer are used for TLC enantiomer separations (e.g. amino acids, thiazolidine derivatives, dipeptides, lactones, α-hydroxycarboxylic acids).
- Chiral separation based on ligand exchange
- Base material is reversed phase nano silica
Impregnation is with Cu(II) ions and as chiral selector a proline derivative.
Customers Who Bought This Also Bought