Featherweight Forceps Set
Catalog #: 470315-238
Supplier: DR INSTRUMENTS INC. SE
Specifications
- Description:Featherweight Forceps Set
- Cat. no.:470315-238
Specifications
About this item
Use a soft touch for delicate specimens.
- Light closure for delicate specimens
Featherweight entomology forceps set with one narrow tip and one wide tip forceps. Made out of flexible stainless steel (Spring Stainless Steel), these forceps can be bent virtually in any shape and form. These forceps are ideal for handling larvae or eggs without crushing them and specimens stored in vials.
Delivery: Set includes broad and fine points and plastic storage case.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Customers Who Bought This Also Bought