ADC® System 5 Portable 5 Cuff Sphygmomanometer
Catalog #: 470315-704
Supplier: AMERICAN DIAGNOSTIC CORP CP
Specifications
- Color:Multiple
- Description:System 5
- Cat. no.:470315-704
Specifications
About this item
The most compact five cuff system.
- Compact enough to fit most trauma kits
- Single connection palm aneroid
- Not made with natural rubber latex
If you need to quickly change cuffs, this kit is a great option. Each cuff has the features you would expect from ADC®. Each cuff in this set is a different color making identification easier, so the proper size can be used. Cuffs are manufactured of high quality denier nylon, and fastening hook and loop adhesive is tested for a minimum of 30000 open and close cycles.
The palm aneroid connects to each cuff using a single tube, bayonet connection, for fast and accurate changes. The manometer operates easily with a single hand, with an ambidextrous release valve trigger. All of this is easily stored in a compact (10×9×4"), dual zippered case with labeled pockets for each component.
Delivery: Includes five sizes of cuffs (child, small adult, adult, large adult and thigh), you should always have a size you need at hand.
Customers Who Bought This Also Bought