ADC® Multikuf 3 Cuff Sphygmomanometer
Catalog #: 470315-702
Supplier: AMERICAN DIAGNOSTIC CORP CP
Specifications
- Color:Black
- Description:Portable Multicuff Kit
- Cat. no.:470315-702
Specifications
About this item
Multiple cuff access in a compact case.
- Durable palm aneroid
- Compact storage case
- Not made with natural rubber latex
If you want easy flexibility in a multi-cuff set, this is a great option. The ADC® 804 palm aneroid is a precision crafted gauge in a rugged ABS housing. To further protect the instrument, it is fully contained in a zinc safety cage. The release valve is an ambidextrous trigger, so its easy to use from either hand, even while wearing gloves. Each cuff is stored in an individually labeled pocket for quick access. The folding pockets, and palm aneroid, are easily stored in a zippered case ( 8×6×3.5") for instant accessibility.
Delivery: The set includes three sizes of adult sized, single tube, cuffs (small adult, adult and large adult) which quickly connect to the aneroid with a bayonet connection.
Customers Who Bought This Also Bought