Entomology Forceps Set
Catalog #: 470315-236
Supplier: DR INSTRUMENTS INC. SE
Specifications
- Description:Entomology Forceps Set
- Material:Stainless Steel
- Cat. no.:470315-236
Specifications
About this item
Designed specifically for the collector.
- For lab or field work
- Includes five different styles of forceps
- Storage pouch
No matter where you do your work, these forceps will help you maintain your collection. Two featherweight forceps (broad and fine point) allow you to handle larvae, eggs, or delicate specimens without damage. A set of pinning forceps with angled tips will make mounting and tagging easy. Finally two microdissection forceps, with ultrafine points, will make delicate manipulations as easy as possible.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Customers Who Bought This Also Bought