Microdissection Forceps Set
Catalog #: 470315-242
Supplier: DR INSTRUMENTS INC. SE
Specifications
- Description:Microdissection Forceps Set
- Material:Stainless Steel
- Cat. no.:470315-242
Specifications
About this item
Precision cut points.
- Includes five varied tip 4.5 in forceps
- Extra fine points
- Storage case
Micro-dissection forceps set contains widely used five pairs of forceps. Each pair of forceps have highly precise and extra fine tips (points) to perform micro-dissection and or any fine work under the microscope. This kit includes two curved and three straight micro-dissection forceps
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Customers Who Bought This Also Bought