ADC® Adscope® 608 Convertible Clinician Stethoscopes
Catalog #: 470315-680
Supplier: AMERICAN DIAGNOSTIC CORP CP
Specifications
- Color:Black
- Description:Adscope 608
- Cat. no.:470315-680
Specifications
About this item
Easily converted from adult to pediatric use..
- Non chill diaphragm and bell rims
- Accessory kit with replacement adsoft ear tips, and an ID tag
- Not made with natural rubber latex
The Adscope 608 is an extremely versatile clinician stethoscope. Adjustable frequency technology (AFD) allow you to tune the diaphragm to enhance high and low frequency response in both adult and pediatric diaphragms. You can convert the pediatric diaphragm to an extra deep bell for unsurpassed low frequency response. The clinician headset, designed to withstand thousands of cycles, features a double leaf tensioning spring and a reinforcing yoke molded into a flexible 22" PVC tubing.
Customers Who Bought This Also Bought