ADC® Proscope 5210 Standard Oto/Ophthalmoscope Set
Catalog #: 470315-660
Supplier: AMERICAN DIAGNOSTIC CORP CP
Specifications
- Description:Proscope 5210 Set
- Cat. no.:470315-660
- Voltage:2.5 V
Specifications
About this item
Competitively priced full size diagnostic set.
- Full sized c cell handle
- Interchangeable oto / ophthalmoscope heads
- Includes 2.5 V Xenon bulb
- Zippered case with fitted insert for storage and protection
This introductory set is ideal for students just staring out. All parts can be stored in a zippered case, which will protect the instruments while bouncing around in a bag. A single C-Cell sized handle easily swaps between the otoscope head and ophthalmoscope head as needed, disposable and reusable specula both have storage areas, and an adaptor allows use of Welch-Allyn specula if needed. Both heads fit to the handle using a bayonet locking mount for a secure connection. The otoscope has a 3X magnifier, as well as an insufflator fitting (bulb and connection not included). The Ophthalmoscope has a lens wheel of 24 corrective lenses from −25 to +40 diopters.
BodyTextAccessories: Does not include batteries.
Customers Who Bought This Also Bought