VWR® Upright Freezer Drawer Rack for 96-Well Microtube Boxes
Catalog #: 76051-748
Supplier: VWR International
Specifications
- Capacity:9
- Length:1015/16"
- Width:57/16"
- Height:75/16"
- Cat. no.:76051-748
Specifications
About this item
Designed for use with 96-well microtube boxes.
- Made of corrosion resistant stainless steel
- Security lock devices are available for each rack
Drawer racks allow easy access to fewer microtiter plates at a time thus protecting other samples in the rack from warming up.
Customers Who Bought This Also Bought