ADC® EKG Ruler
Catalog #: 470315-770
Supplier: AMERICAN DIAGNOSTIC CORP CP
Specifications
- Description:EKG ruler
- Cat. no.:470315-770
Specifications
About this item
A powerful addition for EKG measurement.
- Seconds scale
- Offers both one and two cycle scales
- Small enough to fit in a medical coat pocket
- Complete with protective sleeve
Quickly and easily determine rates from an EKG with this convenient tool. The seconds scale is readily used for PR, QRS, cycle, length, coupling, escape and parasystalic intervals. Easily protected and stored in a clear sleeve, don’t need to worry about spills or splashes.
Customers Who Bought This Also Bought