To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Format:Spin column
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Larger vectors (Cosmid, BAC, PAC, Fosmid)
- Sample type:Bacteria
- Target molecule:Plasmid
- Description:ZR BAC DNA miniprep kit
- Size:100 preps
- Description:ZR BAC DNA miniprep kit
- Yield:Up to 10 µg per preparation, depending on the plasmid copy number, initial volume of E. Coli culture processed, and culture growth conditions. The typical yield is 1 µg per 3 ml of culture.
- Cat. no.:76211-508
Specifications
About this item
The ZR BAC DNA miniprep kit is for efficient isolation of BAC plasmid DNA and other large plasmids (e.g., PAC) from E. coli cell lysates.
- Simple
- Rapid
- User-friendly
- Reliable
- No messy precipitations
Unique column design offers zero buffer retention and low-volume (10 µl) elution. It features a modified alkaline lysis protocol together with unique spin column matrix technology to yield high quality plasmid DNA in minutes and color-coded reagents that allow easy visualization and assessment of complete bacterial cell lysis and buffer neutralization.
The innovative Zymo-Spin™ IC-XL columns are optimized for high yield endotoxin-free plasmid DNA recovery. BAC DNA purified using the ZR BAC DNA miniprep kit is well suited for sequencing, PCR, endonuclease digestion, and more.