ContactUsLinkComponent
Vesphene® III se Disinfectant Cleaner, STERIS®
Vesphene® III se Disinfectant Cleaner, STERIS®
Catalog #: 76303-152
Supplier:  Steris
Vesphene Iii Se Disinfectant 1 Gal CS4
Vesphene® III se Disinfectant Cleaner, STERIS®
Catalog #: 76303-152
Supplier:  Steris
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Vesphene III SE Phenolic Disinfectant
  • Size:
    4×1 gal.
Specifications
About this item
Vesphene III se disinfectant phenolic disinfectant is a one-step, alkaline phenolic cleaner and detergent which is specifically formulated for use on hard, non-porous surfaces.
  • Efficacy - Phenolic active ingredients provide broad spectrum antimicrobial efficacy
  • Productivity - Formulation includes surfactants for surface cleaning while disinfecting
  • Flexibility - Ideal for use in a contamination control rotation
  • Reliability - Complete lot traceability
  • Assurance - Each lot documented by certificate of analysis
Vesphene® III st have dilution rate of 0.8% v/v (1:128), kills a broad spectrum of common bacteria and fungi.
The use-dilution can be applied manually or sprayed.
Customers Who Bought This Also Bought