2,6-Di-tert-butyl-p-cresol ≥99.0% (by GC) E321, SAFC®
Supplier: MilliporeSigma
Certificates
About this item
Synonyms: Butylated hydroxytoluene,Butylhydroxytoluene,2,6-Di-tert-butyl-4-methylphenol
Formula:C₁₅H₂₄O
MW:220.35 g/mol
Boiling Pt:265 °C (1013 hPa)
Melting Pt:69…70 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:127 °C
MDL Number:MFCD00011644
CAS Number:128-37-0
ADR:9,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
:Boiling Pt:
:Boiling Pt: 265 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 69…70 °C
:Flash Pt:
:Flash Pt: 127 °C
:M.W. 220.35 g/mol
:265 °C (1013 hPa)
UN:3077
:9
:III
:127 °C
:GHS09
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (GC):≥99.0%
- Assay (HPLC):99.0 - 101.5%
- Identity (Identification 1 (JPE)):Passes Test
- Identity (IR-spectrum):Passes Test
- Identity (Identification 2 (JPE)):Passes Test
- Identity (HPLC):Passes Test
- Appearance:White to Yellowish Crystalline Powder
- Appearance of solution (100 g/l, Methanol (Ph. Eur.)):Clear and not more intense in color than reference solutionY₅ or BY₅
- Appearance of solution (100 g/l, Ethanol (95 %) (JPE)):Clear and Colorless
- Solidification Temperature:69.2 - 70.0 °C
- Melting Point:69.5 - 72.0 °C
- Absorption Maximum λmax. (Ethanol abs.):277 - 279 nm
- Spec. Absorptivity A 1%/1cm (λmax; 0.02 g/l; Ethanol abs.):81 - 88
- Spec. Absorptivity A 1%/1cm (λ₂₇₈ nm; 0.05 g/l; Ethanol (95 %)):82 - 88
- Heavy Metals (as Pb):≤10 ppm
- Sulfate (SO₄):≤150 ppm
- As (Arsenic):≤3 ppm
- Hg (Mercury):≤1 ppm
- Pb (Lead):≤2 ppm
- tert-Butanol (GC):≤1000 ppm
- Diisobutylene (GC):≤100 ppm
- p-Cresol:≤0.1%
- 2-Propanol (GC):≤2000 ppm
- Related Substances (TLC):≤0.5%
- Related Substances (HPLC) (p-Cresol oder m-cresol):≤0.1%
- Related Substances (HPLC) (3-tert-Butyl-4-hydroxyanisole):≤0.1%
- Related Substances (HPLC) (3,5-Di-tert-butyl-4-hydroxybenzoic acid):≤0.1%
- Related Substances (HPLC) (2-tert-Butyl-4-methylphenol or 2-tert-butyl-5-methylphenol):≤0.1%
- Related Substances (HPLC) (3,5-Di-tert-butyl-4-hydroxy benzaldehyde):≤0.1%
- Related Substances (HPLC) (4,6-Di-tert-butyl-m-cresol):≤0.1%
- Related Substances (HPLC) (2,6-Di-tert-butyl-phenol):≤0.1%
- Related Substances (HPLC) (Any Unspecified Impurity):≤0.1%
- Related Substances (HPLC) (Sum of All Impurities):≤0.7%
- Other Residual Solvents (ICH Q3C):Excluded by the Production Process
- Sulfated Ash (600 °C):≤0.002%
- Water (According to Karl Fischer):≤0.2%
- Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).:
- Meets analytical specifications of Ph Eur, NF, JPE.:
- Conforms to the purity criteria on food additives according to the current European Commission Regulation.:
Specification Test Results
Customers Who Bought This Also Bought