Triacetin (glycerol triacetate) 98.5-100.5% (by alkalimetry), EMPROVE® ESSENTIAL Ph. Eur., BP, USP
Catalog #: CA1.03000.9025
Supplier: MilliporeSigma
CAS Number: 102-76-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Stainless steel drum
- Size:25 L
- Cat. no.:CA1.03000.9025
- Supplier No.:1.03000.9025
Specifications
Specification Test Results
- Assay (Alkalimetric, Calculated on Anhydrous Substance):98.5 - 100.5%
- Assay (GC, calc. on anhydrous substance):97.0 - 102.0%
- Identity (IR-spectrum):Passes Test
- Identity (GC):Passes Test
- Appearance:Clear Colorless, Slightly Viscous Liquid, not more Intense in Color than Reference Solution Y₆
- Acidity (Ph. Eur./ USP):passes test
- Refractive Index (n 20/D):1.430 - 1.432
- Density (d 20/20):1.159 - 1.164
- As (Arsenic):≤1.5 ppm
- Acetic Acid (GC):≤0.1%
- Glycerol (GC):≤0.1%
- 1-Monoacetin (GC):≤0.1%
- 2-Monoacetin (GC):≤0.1%
- 1,3-Diacetin (GC):≤0.1%
- 1,2-Diacetin (GC):≤0.1%
- Individual, Unspecified Impurity (GC):≤0.1%
- Sum of all impurities (GC):≤0.2%
- Acetic Acid (Acidity):≤5000 ppm
- Butylacetate (HS-GC):≤5000 ppm
- Other Residual Solvents (ICH Q3C):Excluded by Manufacturing Process
- Water (According to Karl Fischer):≤0.2%
- Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).:
- Corresponds to Ph. Eur., BP, USP:
Specification Test Results
About this item
Synonyms: Glyceryl triacetate,Propane-1,2,3-triyl triacetate
Boiling Pt:258 °C (1013 hPa)
Melting Pt:–78 °C
Density:1.16 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,5-100,5
Flash Pt:138 °C
:Boiling Pt:
:Boiling Pt: 258 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –78 °C
:Flash Pt:
:Flash Pt: 138 °C
:M.W. 218.21 g/mol
Formula:C₉H₁₄O₆
MW:218.21 g/mol
EINECS:203-051-9
:258 °C (1013 hPa)
:138 °C
:12,09721
MDL Number:MFCD00008716
CAS Number:102-76-1
Customers Who Bought This Also Bought