IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99.0%, Ultra Pure Grade
Supplier: VWR
Certificates
About this item
Synonyms: (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol
Formula:C₉H₁₈O₅S
MW:238.31 g/mol
Boiling Pt:402 °C (1013 hPa)
Melting Pt:109,5…110,5 °C
MDL Number:MFCD00063273
CAS Number:367-93-1
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
:Boiling Pt:
:Boiling Pt: 402 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 109,5…110,5 °C
:Flash Pt:
:Flash Pt:
:M.W. 238.31 g/mol
Storage Temperature:Freezer
EINECS:206-703-0
:402 °C (1013 hPa)
Specification Test Results
- Abs.@300nm (5%, Water):≤ 0.13
- Abs.@400nm (5%, Water):≤ 0.06
- Dioxane Test:PASS / FAIL
- Identification (IR):PASS / Fail
- Melting Point:110 - 114 Degrees C
- Moisture (KF):≤ 1.0 %
- pH (5%, Water) @25C:5 - 7
- Purity:≥ 99.0 %
- Solubility (5%, Water):PASS / Fail
- Specific Rotation (1%, Water):-34.0 - -29.0 degrees
- IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99.0%, Ultra Pure Grade:
Specification Test Results
Related Information
Certificates
Search by Lot Numbersds_VWRV0487
SDS
VWR Life Science Biochemical and Molecular Biology Reagents
Product Literature
Customers Who Bought This Also Bought