Bottles, Stainless Steel, Eagle Stainless
Supplier: Eagle Stainless Container
Certificates
About this item
These STB series bottles are designed for stability studies where storage of the sample in stainless steel is required. Its radiused interior increases its cleanability and optional fill, vent tubes may be added to the endcap.
- 316L stainless steel bottle and endcap
- Machined from certified solid bar 316L stainless steel
- 10 Ra electropolished interior finish
- Traceability/certifications for all stainless-steel lots
- Heat/lot numbers are laser engraved on each bottle
- Air/water-tight design
- Type III DMF
Features 316L endcap, 304 two-segment heavy-duty clamp and platinum-cured silicone gasket (standard).
Customers Who Bought This Also Bought