GTS Express Shipper, Cold Chain Technologies
Catalog #: 10861-124
Supplier: Cold Chain Technologies
Specifications
- Description:GTS Express shipper
- Payload Dimensions:7×5½×3½"
- Cat. no.:10861-124
- Exterior Dimensions:111/4×111/4×65/8"
- Supplier No.:GTS-18-RFG
Specifications
About this item
Express shipper with 2.2 liter payload capacity.
- Pre-qualified for refrigerated (2 to 8°C) for 28 to 36 hours
- Powered by Koolit® advanced PCM
- Specially sized for a 30 day supply of most specialty therapies
- Simple universal pack-out for year-round performance in extreme temperatures
- Lightweight, efficient, freight-saving design minimizes shipping costs
- Stores flat while easy to set up, with a simple, fast pack-out
- Made from environmentally safe and reusable materials
Recommended for use for next day shipments of small size payloads, direct to patient and physician shipments, samples, clinical trials, and diagnostics.
Customers Who Bought This Also Bought