Protective Double Layer Heavy-Duty Aprons, HexArmor
Supplier: HexArmor
Certificates
About this item
The most innovative solution in cut and puncture resistance. Heavy-duty with double layer SuperFabric® brand material. Lightweight, breathable, and ergonomic to ensure safety and comfort.
- Made with SuperFabric® brand material for the highest cut-resistance available in the industry, ANSI/ISEA level A9 (in noted enhanced areas)
- Innovative industrial puncture protection on enhanced areas from wires, metal, wood, and glass
- Heavy-duty (double layer) construction
- Lightweight compared to alternatives
- Launderable
Eliminate snags, punctures, abrasions, or cuts to the arms and torso. HexArmor® industrial aprons and body protection outperform, outlast, and fit better than the competition. SuperFabric® technology provides industry leading cut resistance, providing the protection needed most, where it's needed.
Certifications: EN 388 4X44F, ANSI/ISEA A9.
Specifications
- Catalog No:
- 76208-106
- 76208-108
- 76208-122
- 76208-124
- Garment application:
- Industrial
- Industrial
- Industrial
- Industrial
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Closure type:
- Back fastening
- Back fastening
- Back fastening
- Back fastening
- Fabric:
- SuperFabric®
- SuperFabric®
- SuperFabric®
- SuperFabric®