KoolTemp® Insulated Envelope, Cold Chain Technologies
Catalog #: 80079-854
Supplier: Cold Chain Technologies
Small Business Enterprise
Specifications
- Description:KoolTemp® Insulated Envelope
- Cat. no.:80079-854
- Dimensions:12×16"
- Supplier No.:IE1216-V
Specifications
About this item
KoolTemp® insulated envelopes are lightweight, freight saving, durable, and designed with a spacious payload chamber for temperature-sensitive needs.
- Lightweight, precision-cut expanded insulation
- Spacious, crush-resistant, virtually airtight chamber
- Ideal for personal transportation of medical samples, food, perishables or valuables
- Seal to seal dimensions: 12×16"
An insulated envelope addresses the challenge of heat loss with efficient insulation, reflective exterior surface and a virtually airtight payload chamber.
Customers Who Bought This Also Bought