Notrax® 562 Sanitop® Floor Matting, Justrite®
Catalog #: 76310-654
Supplier: SUPERIOR MANUFACTURING GROUP INC
Specifications
- Color:Black
- Material:Rubber
- Thickness:13 mm (½")
- Length:152 cm (5 ft.)
- Weight:27 lbs.
- Width:91 cm (3 ft.)
Specifications
About this item
Sanitop® mats are made from a durable rubber compound designed to withstand harsh chemicals while creating a slip resistant surface.
- Durable rubber compound
- Molded alternating large/small hole drainage system keeps moisture and debris free from work area
- Raised pattern surrounds large holes to channel drainage and provide worker traction
- Beveled ramps on all 4 sides
- 15 sq. ft. coverage area
A large and small hole drainage system channels liquid and debris away from the work area. Molded-in beveled edges on all four sides eliminate trip hazards. Sanitop® offers a simple, economical solution for wet environments in many industrial or food service applications.
Customers Who Bought This Also Bought