VWR® Horizontal MAXI M Gel Electrophoresis System
Catalog #: 76314-680
Supplier: VWR International
Specifications
- Gel size (W×L):20×25 cm
- Description:VWR® MAXI M Horizontal Gel System
- Cat. no.:76314-680
- L×W×H:290×375×105 mm
- Buffer Volume:2300 ml
Specifications
About this item
Delivery: Includes safety lid with retractable sheathed leads, UVT gel tray with gasketed end gates, three combs - one each 16 well, 24 well and 36 well (1.5 mm thick).
Customers Who Bought This Also Bought