VWR® MINI LR Horizontal Gel Electrophoresis System
Catalog #: 76314-796
Supplier: VWR International
Specifications
- Gel size (W×L):12×14 cm
- Description:VWR® MINI LR Horizontal Gel System
- No. of Samples:8 - 100
- Cat. no.:76314-796
- Buffer Volume:1000 ml
Specifications
About this item
The MINI LR model includes a buffer circulation system, powered by the electrodes themselves.
In constrast to all the other Mini gel systems, the MINI LR model includes a buffer circulation system, powered by the electrodes themselves. Hydrogen bubbles produced at the cathode pass up a diagonally positioned tube connected to the two sides of the tank producing a gentle circulation of the buffer. The result is a no pH and ionic gradient - without the need of mechanical pumps.
Delivery: Includes integral buffer circulation, safety lid with retractable sheathed leads, UVT gel tray, two combs, one 12 well and one 20 well (1.5 mm thick).
Customers Who Bought This Also Bought