Magnesium sulfate, anhydrous 99.0-100.5% (excluding ignited substances), powder USP, Macron Fine Chemicals™
Catalog #: CAMK5054-26
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 7487-88-9
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:200 lbs.
- Cat. no.:CAMK5054-26
Specifications
Specification Test Results
- Meets U.S.P Requirements:
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- Appearance:Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Loss on Drying at 105°C:≤ 2 %
- Loss on Ignition:≤ 2.0 %
- USP - pH (1 in 20):5.0 - 9.2
- USP - Chloride (Cl):≤ 0.014 %
- USP - Selenium (Se):≤ 0.003 %
- USP - Iron (Fe):≤ 20 ppm
- USP - Assay (as MgSO4) (ignited basis):99.0 - 100.5 %
- Solubility:Passes Test
- Thru U.S. No. 100 Sieve:≥ 85 %
- Appearance (fine, white powder):
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on:
- elemental impurities for this product is available on the:
- associated Product Regulatory Data Sheet and elemental impurity:
- profile report.:
Specification Test Results
About this item
Synonyms: Sulfuric acid magnesium salt (1:1),Epsom salt,Magnesium(II) sulfate
:Storage Temperature: Ambient
Melting Pt:1127 °C
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 1127 °C
:Flash Pt:
:Flash Pt:
:M.W. 120.37 g/mol
:Magnesium sulfate
MW:120.37 g/mol
Storage Temperature:Ambient
:M.O.C. Category R
:12,05731
Formula:MgSO₄
MDL Number:MFCD00011110
CAS Number:7487-88-9
Customers Who Bought This Also Bought