Flash Western Cheminuminescence Kit, Azure Biosystems
Supplier: Azure Biosystems
Certificates
About this item
The flash western chemilumnescence kit allows to run a rapid and sensitive chemiluminescence western blot. Block, hybridize, enhance signal, and detect in less than one hour of blotting time.
- Fast protocol with few hands on steps
- Antibody stabilization for enhanced chemi signal
- Low background and increased signal to noise ratios
- Increased signal detection and linear dynamic range
Delivery: Kit includes 10 PVDF membranes, chemi blot blocking buffer (100 ml), flash HRP conjugate (500 μl), flash wash buffer (250 ml) and radiance ECL substrate (100 ml).
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.