Order Entry
ContactUsLinkComponent
Serological Pipettes with Cotton Plugging Top, Bellco Glass
Serological Pipettes with Cotton Plugging Top, Bellco Glass
Catalog #: CA12106-810
Supplier:  Bellco Glass
PIPET, GLASS 50 ML CS12
Serological Pipettes with Cotton Plugging Top, Bellco Glass
Catalog #: CA12106-810
Supplier:  Bellco Glass

Specifications

  • Size:
    50×0.2 ml
  • Cat. no.:
    CA12106-810
  • Color Code:
    Red
  • Tolerance:
    ±0.4 ml

Specifications

About this item

Type 1 Class B Borosilicate Glass.

  • Calibrated to tip
  • Tempered tip, to deliver, blow out

Constricted top for cotton plugging.

Delivery: Supplied without cotton.
Customers Who Bought This Also Bought