Horizon Wind to Hydrogen Science Kit
Catalog #: 470314-468
Supplier: HORIZON FUEL CELL TECHNOLOGIES
Specifications
- Description:Wind to Hydrogen Science Kit
- Cat. no.:470314-468
Specifications
About this item
Demonstrate the principles of wind energy, hydrogen fuel cells, electrolysis and basic electronics.
- Turn wind power into hydrogen energy
- Build your own hydrogen fuel cell powered inventions
- Features revolutionary new Horizon wind turbine
- Includes Horizon’s complete curriculum on renewable energy on CD
The Wind to Hydrogen Science Education Kit enables students to invent their own clean energy applications using a small electric motor powered by a fuel cell. Hydrogen is produced through water electrolysis by a reversible PEM fuel cell that is itself powered by a wind turbine with profiled blades based on NASA aeronautics. This hydrogen is then converted to electrical energy by the fuel cell and can be used to power a small fan.
Customers Who Bought This Also Bought