Horizon Eneregy Monitor
Catalog #: 470314-474
Supplier: HORIZON FUEL CELL TECHNOLOGIES
Specifications
- Description:Data Logging Energy Monitor
- Cat. no.:470314-474
Specifications
About this item
Compatible with any of Horizon's science education kits.
- Windows and mac compatible
- Use with or without a computer
- Export data to excel or CSV format
- Record videos for playback and review
Voltage, current, power, joules, resistance and even RPM speed available at the touch of a button—displayed either on the LCD screen of the monitor itself or as graphical data on your PC. The Horizon Energy Monitor helps bring lab and field experiments back into the classroom, making it the ideal tool to complement the wind to hydrogen science Kit (FCJJ-56), the renewable energy science kit (FCJJ-37) and the wind energy science kit (FCJJ-39).
Certifications: CoC, RoHS, EN 71:PART1;PART2;PART3, EN 62115, PHTH-EU, ASTM F963, CPSIA-LEAD, CPSIA-PHTHALATES, REACH, CA
Customers Who Bought This Also Bought