Trifluoroacetic acid ≥99.0% (by GC), BioSyn™ for biosynthesis, for HPLC, Burdick & Jackson™
Catalog #: CABJ91707250ML
Supplier: Burdick & Jackson
New Product
CAS Number: 76-05-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle
- Size:250 ml
Specifications
Specification Test Results
- Refractive Index:n20/D 1.3 (lit.)
- Assay (GC):Min. 99.0 %
- Suitability test (HPLC):complying
- Water (Karl Fischer):Max. 0.05 %
- Absorbance at 320 nm:Max. 0.025
- Absorbance at 300 nm:Max. 0.03
- Absorbance at 290 nm:Max. 0.04
- Absorbance at 280 nm:Max. 0.05
- Absorbance at 270 nm:Max. 0.1
- Absorbance at 260 nm:Max. 0.9
- APHA:Max. 25
- Burdick & Jackson™, Trifluoroacetic acid ≥99.0% (by GC), BioSyn™ for biosynthesis, for HPLC:
Specification Test Results
About this item
Synonyms: TFA
Boiling Pt:72 °C (1.013 hPa)
Melting Pt:-15 °C
Density:1.48 g/cm³ (20 °C)
ADR:8,I
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
Flash Pt:100 °C
:Boiling Pt:
:Boiling Pt: 72 °C (1.013 hPa)
:Melting Pt:
:Melting Pt: -15 °C
:Flash Pt:
:Flash Pt: 100 °C
:M.W. 114.02 g/mol
Formula:C₂HF₃O₂
MW:114.02 g/mol
MDL Number:MFCD00004169
CAS Number:76-05-1
EINECS:200-929-3
:72 °C (1.013 hPa)
Storage Temperature:Ambient
UN:2699
:8
:I
:100 °C
:GHS07
:GHS05
:12,09812
Customers Who Bought This Also Bought