8100 Series Poly Shell Stacking Chairs, National Public Seating
Supplier: National Public Seating
Certificates
About this item
The lightweight 8100 Poly Shell series is ideal for environments such as cafeterias and lounges.
- One piece, injection molded, color-impregnated, high-density polypropylene seat/back
- Anti-static shell and frame shields to protect against snags and pinching
- Frame is chrome plated with 17-gauge 7/8" square tubing
- Five underseat cross braces for added stability and durability
- 6 stack bumpers on each chair protect finish
Can be stacked 12 high
10 year warranty
Certifications: MAS Certified Green, ANSI BIFMA.
Specifications
- Catalog No:
- 76276-688
- 76276-488
- 76276-684
- 76276-692
- Model:
- 76276-688
- 76276-488
- 76276-684
- 76276-692
- Frame colour/finish:
- Chrome
- Chrome
- Chrome
- Chrome
- Seat/back color:
- Blue
- White
- Black
- Light oak
- Seat/back material:
- PP
- PP
- PP
- PP
- Seat dimensions:
- W17×D16.75×H18"
- W18.25×H8.75"
- W17×D16.75×H18"
- W18.25×H8.75"
Specifications
Customers Who Bought This Also Bought